Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clostridium Difficile Toxin A Antibody (PCG4.1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 18 publications
Supplier: Novus Biologicals NB60010660.1MG
Description
Clostridium Difficile Toxin A Monoclonal specifically detects Clostridium Difficile Toxin A in Bacteria samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Clostridium Difficile Toxin A | |
Monoclonal | |
1.16 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
tcdA, toxA | |
Full length protein A (C. difficile). | |
0.1 mg | |
Immunology | |
C. difficile Toxin A. Does not cross react with C. difficile Toxin B | |
Bacteria, Hamster | |
Purified |
Western Blot, ELISA | |
PCG4.1 | |
Unconjugated | |
10mM Sodium Phosphate (pH 7.4) and 0.15M NaCl with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction