Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clostridium difficile Toxin A Chicken anti-Bacteria, HRP, Polyclonal, Gallus

Chicken Polyclonal Antibody
Supplier: Vector Laboratories ACDTAHRP

This item is not returnable.
View return policy
Description
Clostridium difficile Toxin A Polyclonal antibody specifically detects Clostridium difficile Toxin A in Bacteria samples. It is validated for ELISA, Western BlotSpecifications
Clostridium difficile Toxin A | |
Polyclonal | |
HRP | |
TcdA | |
Clostridium difficile Toxin A | |
0.1 mg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.5-2 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH | |
Chicken | |
Affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgY |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction