Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clostridium difficile Toxin A Monoclonal Antibody (A73H)
GREENER_CHOICE

Catalog No. PIMA518192
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518192 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518192 Supplier Invitrogen™ Supplier No. MA518192
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not cross-react with Toxin B. Purity is >90%. Recommended pairs for ELISA: MA5-18192 (capture) and MA5-18191 (conjugated, detection).

Clostridium difficile is a species of bacteria of the genus Clostridium which are Gram-positive, anaerobic, spore-forming rods (bacillus). C.difficile is the most significant cause of pseudomembranous colitis, a severe infection of the colon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridium difficile Toxin A
Applications ELISA
Classification Monoclonal
Clone A73H
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias TcdA
Host Species Mouse
Immunogen Cl. difficile native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.