Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Clostridium difficile Toxin A Monoclonal Antibody (L0K)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518191
Description
This antibody does not cross-react with Toxin B. Purity is >90%. Recommended pairs for ELISA: MA5-18192 (capture) and MA5-18191 (conjugated, detection).
Clostridium difficile is a species of bacteria of the genus Clostridium which are Gram-positive, anaerobic, spore-forming rods (bacillus). C.difficile is the most significant cause of pseudomembranous colitis, a severe infection of the colon.
Specifications
Clostridium difficile Toxin A | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG2a |
ELISA | |
L0K | |
Unconjugated | |
TcdA | |
Cl. difficile native protein. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction