Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clostridium difficile Toxin A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA185042
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA185042 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA185042 Supplier Invitrogen™ Supplier No. PA185042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabbit anti Clostridium difficile toxin A antibody recognizes Clostridium difficile toxin A.

Clostridium difficile is a species of bacteria of the genus Clostridium which are Gram-positive, anaerobic, spore-forming rods (bacillus). C.difficile is the most significant cause of pseudomembranous colitis, a severe infection of the colon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridium difficile Toxin A
Applications ELISA
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias TcdA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the amino acids 2123 -2132 of the native molecule conjugated to Keyhole limpet Haemocyanin. Peptide sequence: CTIDGKKYYFN.
Purification Method Protein G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.