Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium Perfringens Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10062474 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062474 0.5 mL
NB017482 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062474 Supplier Novus Biologicals Supplier No. NB10062474
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Clostridium Perfringens Polyclonal specifically detects Clostridium Perfringens in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Clostridium Perfringens
Applications ELISA
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:5000-1:20000
Formulation Phosphate Buffered Saline with 0.09% Sodium Azide
Host Species Rabbit
Immunogen Clostridium perfringens ATCC 13124
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CLOSTRIDIUM PERFRINGENS
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.