Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clostridium perfringens Neuraminidase Recombinant Rabbit Monoclonal Antibody (112), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543055
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543055 100 μL
PIMA543473 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543055 Supplier Invitrogen™ Supplier No. MA543055
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Clostridium Perfringens Neuraminidase

Sialidase, also known as Neuraminidase, is a secreted protein from bacterial species Clostridium perfringens that catalyses hydrolysis of glycosidic linkages on sialic acid residues in many biomolecules. It is suggested to be involved in the pathogenesis of microbial infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridium perfringens Neuraminidase
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 112
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Sialidase
Host Species Rabbit
Immunogen Recombinant Clostridium Perfringens Neuraminidase protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.