Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clostridium Tetani Toxoid Antibody (6E7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$418.55
Specifications
| Antigen | Clostridium Tetani Toxoid |
|---|---|
| Clone | 60000000 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP22355401
![]() |
Novus Biologicals
NBP2235540.1ML |
0.1 mL |
Each of 1 for $418.55
|
|
|||||
Description
Clostridium Tetani Toxoid Monoclonal specifically detects Clostridium Tetani Toxoid in Bacteria samples. It is validated for ELISA.Specifications
| Clostridium Tetani Toxoid | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
| TETX | |
| IgG1 κ | |
| Protein A or G purified | |
| Clone 6E7 binds to both tetanus toxin and tetanus toxoid |
| 60000000 | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Bacteria | |
| Clostridium tetani toxoid, Tentoxylysin, Tetanus toxin, tetX | |
| Tetanus toxoid adsorbed to aluminum hydroxide gel. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title