Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Clostripain (Cl. Histolyticum)

Catalog No. 89939880 Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 mg

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Name Clostripain (Cl. Histolyticum)
Preparation Method Native protein chromatographically purified from Clostridium histolyticum
Quantity 1 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name CLOSTRIPAIN
Assays Clostripain Assay; Peptide mapping; Sequence analysis; Cell isolation; Hydrolysis/condensation of amide bonds; Peptide synthesis.
Biological Activity ≥50 units per mg dry weight. One Unit hydrolyzes one micromole of N-benzoyl-L-arginine ethyl ester per minute at 25°C, pH 7.6, in the presence of dithiothreitol.
Species Clostridium species
Protein Tag None
Expression System Native protein chromatographically purified from Clostridium histolyticum
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.