Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clotrimazole, Powder, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601766
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle

CL127, 23593-75-1, C22H17ClN2

Spectrum™ Chemical Clotrimazole, Powder, USP is an azole antifungal used to treat skin infections. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 23593-75-1
Molecular Formula C22H17ClN2
Molecular Weight (g/mol) 344.84
InChI Key VNFPBHJOKIVQEB-UHFFFAOYSA-N
IUPAC Name 1-[(2-chlorophenyl)diphenylmethyl]-1H-imidazole
SMILES ClC1=CC=CC=C1C(N1C=CN=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98-102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.