Learn More
Science First™ Cloud Chamber
SDPSCI_ED
Catalog No. s52008
Click to view available options
:
Cloud Chamber Kit
Includes
Mail-away coupon for the source of alpha and beta particles
Requires
Liquid nitrogen or dry ice
Study the rate of alpha decay and range of alpha particles.
Tracks of radioactive particles are made visible by the condensation of droplets in a thin layer of alcohol vapor.
Specifications
Product Type Kit, Cloud Chamber
For Use With (Application) Study the rate of alpha decay and range of alpha particles
Includes Mail-away coupon for the source of alpha and beta particles
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.