Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Clozapine N-oxide
GREENER_CHOICESDP

Catalog No. 493650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-365-0 50 mg
49-361-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-365-0 Supplier R&D Systems™ Supplier No. 4936/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Activator of muscarinic DREADDs

Metabolite of clozapine (Cat. No. 0444). Activates human muscarinic designer receptors (DREADDs) in vitro. Silences hippocampal neurons expressing hM4 DREADDs in vitro. Activates Gs-linked DREADDS- AgRP in mice via intracerebroventricular (i.c.v) injection. Shown to be a P-glycoprotein (P-gp) efflux pump substrate. Other potent and selective hM3Dq DREADD agonists available: DREADD agonist 21 and Perlapine .

Water soluble salt also available.

This product contains 1 molar equivalent of ethanol.

Chemical Identifiers

CAS 34233-69-7
Molecular Formula C18H19ClN4O

Specifications

Assay Percent Range >98%
Quantity 50 mg
Product Line Tocris
Formula Weight 342.82
Purity >99%
Molecular Weight (g/mol) 342.827
Chemical Name or Material Clozapine N-oxide
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.