Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CLP 290
GREENER_CHOICE

Catalog No. 675810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-581-0 10 mg
67-585-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-581-0 Supplier Tocris Bioscience™ Supplier No. 6758/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

K+/Cl- cotransporter 2 (KCC2) activator; orally bioavailable prodrug of CLP 257 (Cat. No.5242)

CLP 290 is a K+/Cl- Cotransporter 2 (KCC2) activator. Activates spinal inhibitory interneurons, restores physiological neuronal activity in the spinal cord and induces functional recovery in a mouse model of spinal cord injury. Also decreases blood vasopressin and glucose levels in mice with Streptozotocin (Cat. No. 1621) induced diabetes mellitus. Orally bioavailable prodrug of CLP 257 (Cat. No.5242).

Specifications

Chemical Name or Material (Z)-5-Fluoro-2-((4-oxo-2-(tetrahydropyridazin-1(2 H)-yl)thiazol-5(4 H)-ylidene)methyl)phenyl pyrrolidine-1-carboxylate
CAS 1181083-81-7
Quantity 10 mg
Target KCC2 Activators
Molecular Formula C19H21FN4O3S
Formula Weight 1990
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.