Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLP24/TMEM204 Antibody (652424), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6747F
Description
CLP24/TMEM204 Monoclonal antibody specifically detects CLP24/TMEM204 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
CLP24/TMEM204 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
79652 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunocytochemistry, CyTOF | |
652424 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
C16orf30chromosome 16 open reading frame 30, Claudin-like protein 24, CLP24MGC111564, FLJ20898, transmembrane protein 204 | |
NS0 mouse myeloma cell line transfected with human CLP24, Accession # Q9BSN7 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction