Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLPB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564175
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564175 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564175 Supplier Invitrogen™ Supplier No. PA564175
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRKENGWYDE EHPLVFLFLG SSGIGKTELA KQTAKYMHKD AKKGFIRLDM SEFQERHEVA KFIGSPPGYV GHEEGGQLTK KLKQCPNAVV LF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

CLPB may function as a regulatory ATPase and be related to secretion/protein trafficking process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Clpb
Gene Accession No. Q9H078
Gene Alias AL118244; ANKCLB; ankyrin-repeat containing bacterial clp fusion; caseinolytic mitochondrial matrix peptidase chaperone subunit B; caseinolytic peptidase B; caseinolytic peptidase B protein homolog; CLPB; ClpB caseinolytic peptidase B; ClpB caseinolytic peptidase B homolog; ClpB caseinolytic peptidase B homolog (E. coli); ClpB homolog, mitochondrial AAA ATPase chaperonin; HSP78; MEGCANN; MGCA7; Skd3; suppressor of K+ transport defect 3; suppressor of potassium transport defect 3; testicular secretory protein Li 11
Gene Symbols Clpb
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLPB. Recombinant protein control fragment (Product #RP-104140).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81570
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.