Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLPH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560590 Supplier Invitrogen™ Supplier No. PA560590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTTNSITRDS ITEHFMPVKI GNISSPVTTV SLIDFSTDIA KEDILLATID TGDAEISITS EVSGTLKDSS AGVADAPAFP RKKDEADMSN YNSSIKSNVP Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 59%.

CLPH (Casein-Like phosphoprotein) is a recently discovered, evolutionarily conserved, testis-specific cytoplasmic protein expressed exclusively in postmeiotic germ cells in rat spermatogenesis. CLPH is a novel casein kinase 2-phosphorylated disordered and calcium-binding protein that displays strong variation in relative expression levels during spermatogenesis. It is frequently associated with the mitochondrial inner membrane during the last steps of spermatid differentiation and is involved in the extremely complex structural rearrangements occurring in haploid germ cells during spermiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CABS1
Gene Accession No. Q96KC9
Gene Alias C4orf35; CABS1; calcium binding protein, spermatid associated 1; calcium-binding and spermatid-specific protein 1; calcium-binding protein, spermatid-specific 1; calcium-binding protein, sperm-specific 1; casein-like phosphoprotein; Clph; NYD-SP26; Protein RSD-6; Rsd6; RSD-6; Testis development protein NYD-SP26
Gene Symbols CABS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLPH. Recombinant protein control fragment (Product #RP-100151).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85438
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.