Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLPP Mouse anti-Human, Porcine, Clone: 2E1D9, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 662711IG150UL

Description
The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.Specifications
CLPP | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16740 | |
CLPP | |
CLPP Fusion Protein Ag8420 | |
150 μL | |
Primary | |
Human, Pig | |
Antibody | |
IgG3 |
Immunohistochemistry (Paraffin), Western Blot | |
2E1D9 | |
Unconjugated | |
CLPP | |
CLPP, Endopeptidase Clp | |
Mouse | |
Protein A | |
RUO | |
100737271, 8192 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction