Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLPP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597185
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597185 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597185 Supplier Invitrogen™ Supplier No. PA597185
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLIPIVVEQT GRGERAYDIY SRLLRERIVC VMGPIDDSVA SLVIAQLLFL QSESNKKPIH MYINSPGGVV TAGLAIYDTM QYILNPICTW CVGQAASMGS LLLAAGTPGM RHSLPNSRIM IHQPSGGARG QATDIAIQAE EIMKLKKQLY NIYAKHTKQS LQVIESAMER DRYMSPMEAQ EFGILDKVLV HPPQDGEDEP TLVQKEPVEA APAAEPVPAS T Positive Samples: LO2, A-549, BxPC-3, 22Rv1, Mouse liver, Mouse kidney, Mouse heart, Mouse lung, Rat liver; Cellular Location: Mitochondrion matrix.

The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CLPP
Gene Accession No. O88696, Q16740
Gene Alias ATP-dependent Clp protease proteolytic subunit, mitochondrial; ATP-dependent protease ClpAP, proteolytic subunit, human; AU019820; caseinolytic mitochondrial matrix peptidase proteolytic subunit; caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; caseinolytic protease, ATP-dependent, proteolytic subunit homolog; Clpp; ClpP caseinolytic peptidase ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog; D17Wsu160e; DFNB81; endopeptidase Clp; LOW QUALITY PROTEIN: ATP-dependent Clp protease proteolytic subunit, mitochondrial; PRLTS3; putative ATP-dependent Clp protease proteolytic subunit, mitochondrial
Gene Symbols CLPP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 57-277 of human CLPP (NP_0060031).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301117, 53895, 8192
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.