Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLPS Antibody (111), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290233MFV450
Description
CLPS Monoclonal antibody specifically detects CLPS in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
CLPS | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
111 | |
ELISA, Immunohistochemistry-Paraffin | |
colipase, colipase, pancreatic, pancreatic colipase preproprotein | |
Rabbit immunized with purified, recombinant Human CLPS (Uniprot#: P04118, Met1-Gln112) | |
0.1 mL | |
Lipid and Metabolism | |
1208 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction