Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLPS Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299955H
Description
CLPS Polyclonal specifically detects CLPS in Human samples. It is validated for ELISA.Specifications
CLPS | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
HRP | |
colipase, colipase, pancreatic, pancreatic colipase preproprotein | |
Produced in rabbits immunized with purified, recombinant CLPS (P04118; Met1-Gln112). | |
0.1 mL | |
Lipid and Metabolism | |
1208 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction