Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLPS Monoclonal Antibody (04)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530246
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CLPS.
The protein encoded by this gene is a cofactor needed by pancreatic lipase for efficient dietary lipid hydrolysis. It binds to the C-terminal, non-catalytic domain of lipase, thereby stabilizing an active conformation and considerably increasing the overall hydrophobic binding site. The gene product allows lipase to anchor noncovalently to the surface of lipid micelles, counteracting the destabilizing influence of intestinal bile salts. This cofactor is only expressed in pancreatic acinar cells, suggesting regulation of expression by tissue-specific elements. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
CLPS | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P04118 | |
CLPS | |
Recombinant Human CLPS Protein (Met1-Gln112). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
04 | |
Unconjugated | |
CLPS | |
2200003J09Rik; Clps; COL; Colipase; colipase, pancreatic; COLQ; pancreatic colipase preproprotein | |
Mouse | |
Protein A | |
RUO | |
1208 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction