Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLPS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552699
Description
Immunogen sequence: NLENGELCMN SAQCKSNCCQ HSSALGLARC TSMASENSEC SVKTLYGIYY KCPCERGLTC EGDKTIVGSI TNTNFGICHD AGR Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.
The protein encoded by this gene is a cofactor needed by pancreatic lipase for efficient dietary lipid hydrolysis. It binds to the C-terminal, non-catalytic domain of lipase, thereby stabilizing an active conformation and considerably increasing the overall hydrophobic binding site. The gene product allows lipase to anchor noncovalently to the surface of lipid micelles, counteracting the destabilizing influence of intestinal bile salts. This cofactor is only expressed in pancreatic acinar cells, suggesting regulation of expression by tissue-specific elements. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
CLPS | |
Polyclonal | |
Unconjugated | |
CLPS | |
2200003J09Rik; Clps; COL; Colipase; colipase, pancreatic; COLQ; pancreatic colipase preproprotein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1208 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P04118 | |
CLPS | |
Recombinant protein corresponding to Human CLPS. Recombinant protein control fragment (Product #RP-90264). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction