Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLPS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588748 Supplier Invitrogen™ Supplier No. PA588748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse pancreas, Rat pancreas Immunogen sequence: SVAYAAPGPR GIIINLENGE LCMNSAQCKS NCCQHSSALG LARCTSMASE NSECSVKTLY GIYYKCPCER GLTCEGDKTI VGSITNTNFG ICHDAGRSKQ.

The protein encoded by this gene is a cofactor needed by pancreatic lipase for efficient dietary lipid hydrolysis. It binds to the C-terminal, non-catalytic domain of lipase, thereby stabilizing an active conformation and considerably increasing the overall hydrophobic binding site. The gene product allows lipase to anchor noncovalently to the surface of lipid micelles, counteracting the destabilizing influence of intestinal bile salts. This cofactor is only expressed in pancreatic acinar cells, suggesting regulation of expression by tissue-specific elements. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPS
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CLPS
Gene Accession No. P04118, P17084, Q9CQC2
Gene Alias 2200003J09Rik; Clps; COL; Colipase; colipase, pancreatic; COLQ; pancreatic colipase preproprotein
Gene Symbols CLPS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 13-112 of human CLPS (NP_0018231).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109791, 1208, 25680
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.