Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLPTM1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232114C
Description
CLPTM1 Polyclonal antibody specifically detects CLPTM1 in Human samples. It is validated for Western BlotSpecifications
CLPTM1 | |
Polyclonal | |
Western Blot | |
cleft lip and palate associated transmembrane protein 1, cleft lip and palate transmembrane protein 1 | |
The immunogen this antibody was made to, maps to a region between residue 619 to 669 of human Cleft Lip and Palate Associated Transmembrane Protein 1 using the numbering given in entry NP_001285.1 (GeneID 1209). | |
0.1 mL | |
Cell Biology | |
1209 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction