Learn More
Invitrogen™ CLPTM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596750
Description
Immunogen sequence: MAAAQEADGA RSAVVAAGGG SSGQVTSNGS IGRDPPAETQ PQNPPAQPAP NAWQVIKGVL FRIFIIWAIS SWFRRGPAPQ DQAGPGGAPR VASRNLFPKD TLMNLHVYIS EHEHFTDFNA TSALFWEQHD LVYGDWTSGE NSDGCYEHFA ELDIPQSVQQ NGSIYIHVYF TKSGFHPDPR QKALYRRLAT VHMSRMINKY KRRRFQKTKN LLTGETEADP EMIKRAEDYG PVEVISHWHP NITINIVDDH TPWVKGSVPP PLDQYVKFDA VSGDYYPIIY; Positive Samples: Mouse kidney, Mouse brain, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein.
CLPTM1 may play a role in T-cell development.
Specifications
CLPTM1 | |
Polyclonal | |
Unconjugated | |
CLPTM1 | |
cleft lip and palate associated transmembrane protein 1; cleft lip and palate transmembrane protein 1; Cleft lip and palate transmembrane protein 1 homolog; CLPTM1; CLPTM1, transmembrane protein; HS9; N14; Thymic epithelial cell surface antigen | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1209, 292696, 56457 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O96005, Q8VBZ3 | |
CLPTM1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CLPTM1 (NP_0012851). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.