Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLPTM1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP317231100UL
This item is not returnable.
View return policy
Description
CLPTM1 Polyclonal antibody specifically detects CLPTM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CLPTM1 | |
Polyclonal | |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
cleft lip and palate associated transmembrane protein 1, cleft lip and palate transmembrane protein 1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: FAFVIKMPVMYRIGCLRDDVVFFIYLYQRWIYRVDPTRVNEFGMSGEDPTAAAPVAEVPTAAGALTPTPAPTTTTATREEASTSLPTKPTQ | |
100 μg | |
Cell Biology | |
1209 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction