Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLPTM1L Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299271AF488
Description
CLPTM1L Polyclonal specifically detects CLPTM1L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CLPTM1L | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
cisplatin resistance related protein CRR9p, Cisplatin resistance-related protein 9, cleft lip and palate transmembrane protein 1-like protein, CLPTM1-like, CLPTM1-like protein, CRR9DKFZp761M2324, CRR9p, DKFZp666M1010, FLJ14400, FLJ32533 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human CLPTM1L. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
81037 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction