Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLPTM1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550639
Description
The antibody detects endogenous levels of total CLPTM1L protein.
CLPTM1L enhances cisplatin-mediated apoptosis, when overexpressed.
Specifications
CLPTM1L | |
Polyclonal | |
Unconjugated | |
CLPTM1L | |
C130052I12Rik; cisplatin resistance related protein CRR9p; cisplatin resistance-related protein 9; Cleft lip and palate transmembrane protein 1-like protein; CLPTM1 like; Clptm1l; CLPTM1-like; CLPTM1-like protein; CRR9; CRR9p; RGD1307896 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81037 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96KA5 | |
CLPTM1L | |
Synthetic peptide corresponding to a region derived from internal residues of human CLPTM1-like. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction