Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLPTM1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117703
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117703 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117703 Supplier Invitrogen™ Supplier No. PA5117703
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

CLPTM1L enhances cisplatin-mediated apoptosis, when overexpressed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPTM1L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CLPTM1L
Gene Accession No. Q96KA5
Gene Alias C130052I12Rik; cisplatin resistance related protein CRR9p; cisplatin resistance-related protein 9; Cleft lip and palate transmembrane protein 1-like protein; CLPTM1 like; Clptm1l; CLPTM1-like; CLPTM1-like protein; CRR9; CRR9p; RGD1307896
Gene Symbols CLPTM1L
Host Species Rabbit
Immunogen Recombinant Human Cleft lip and palate transmembrane protein 1-like protein (32-172aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81037
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.