Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLPX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565501 Supplier Invitrogen™ Supplier No. PA565501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIVFLDEVDKI GSVPGIHQLR DVGGEGVQQG LLKLLEGTIV NVPEKNSRKL RGETVQVDTT NILFVASGAF NGLDRIISRR KNEKYLGFGT PS Highest antigen sequence identity to the following orthologs - mouse 99%, rat 100%.

ATP-dependent specificity component of the Clp protease. It directs the protease to specific substrates. Can perform chaperone functions in the absence of clpP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPX
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLPX
Gene Accession No. O76031
Gene Alias ATP-dependent Clp protease ATP-binding subunit clpX-like, mitochondrial; AU014732; caseinolytic mitochondrial matrix peptidase chaperone subunit; caseinolytic peptidase X; caseinolytic protease X; CLPX; ClpX caseinolytic peptidase X homolog; ClpX caseinolytic protease X homolog; E330029I21; energy-dependent regulator of proteolysis
Gene Symbols CLPX
Host Species Rabbit
Immunogen Recombinant Human CLPX. Recombinant protein control fragment (Product #RP-106178).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10845
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.