Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLPX Recombinant Rabbit Monoclonal Antibody (JE62-96)
GREENER_CHOICE

Catalog No. PIMA544792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544792 Supplier Invitrogen™ Supplier No. MA544792
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CLPX Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

ATP-dependent specificity component of the Clp protease. It directs the protease to specific substrates. Can perform chaperone functions in the absence of clpP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPX
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE62-96
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CLPX
Gene Accession No. O76031, Q5U2U0, Q9JHS4
Gene Alias ATP-dependent Clp protease ATP-binding subunit clpX-like, mitochondrial; AU014732; caseinolytic mitochondrial matrix peptidase chaperone subunit; caseinolytic peptidase X; caseinolytic protease X; CLPX; ClpX caseinolytic peptidase X homolog; ClpX caseinolytic protease X homolog; E330029I21; energy-dependent regulator of proteolysis
Gene Symbols CLPX
Host Species Rabbit
Immunogen Synthetic peptide within human CLPX aa 584-633/633.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10845, 270166, 300786
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.