Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clr-a Rat anti-Mouse, Clone: 2000, MilliporeSigma™

Catalog No. MABF220925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF220925 25 μL
MABF2209100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF220925 Supplier MilliporeSigma Supplier No. MABF220925UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Clr-a Monoclonal antibody specifically detects Clr-a in Mouse samples. It is validated for Immunohistochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clr-a
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2000
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rat
Immunogen c-Myc/His-tagged recombinant fragment corresponding to 140 amino acids from the extracellular domain of murine C-type lectin domain family 2 member E (Clr-a).
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.