Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ clr6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144562 Supplier Invitrogen™ Supplier No. PA5144562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

clr6 Polyclonal Antibody for Western Blot, ELISA

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Has a role in chromatin assembly and chromosome segregation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen clr6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene clr6
Gene Accession No. O59702
Gene Alias Histone deacetylase clr6 clr6 SPBC36.05c
Gene Symbols clr6
Host Species Rabbit
Immunogen Recombinant Schizosaccharomyces pombe (strain 972/ATCC 24843) (Fission yeast) clr6 protein (1-239aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2540368
Target Species Yeast
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.