Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLRN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144089
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144089 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144089 Supplier Invitrogen™ Supplier No. PA5144089
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLRN2 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

Clarin 2 belongs to the clarin family of genes. It's a small integral membrane glycoproteins with four transmembrane domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLRN2
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 4.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene CLRN2
Gene Accession No. A0PK11
Gene Alias clarin 2; clarin-2; CLRN2; EG624224
Gene Symbols CLRN2
Host Species Rabbit
Immunogen Recombinant Human Clarin-2 protein (31-100AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645104
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.