Learn More
Invitrogen™ CLRN3 Polyclonal Antibody

Description
CLRN3 Polyclonal Antibody for Flow, Western Blot
Specifications
Specifications
| Antigen | CLRN3 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.37 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CLRN3 |
| Gene Accession No. | Q8NCR9 |
| Gene Alias | clarin 3; clarin-3; CLRN3; TMEM12; transmembrane protein 12; USH3AL1; Usher syndrome type-3A-like protein 1 |
| Gene Symbols | CLRN3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.