Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLSTN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144177
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144177 Supplier Invitrogen™ Supplier No. PA5144177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLSTN2 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

Calsyntenin 2 (CLSTN2) is a gene located on chromosome 3p22.2 that encodes a member of the calsyntenin family of proteins, which are characterized by their role in calcium binding and involvement in synaptic function and neural connectivity. CLSTN2 is predominantly expressed in the nervous system, where it contributes to synaptic organization and signaling processes essential for neural plasticity and cognitive functions. The protein is involved in mediating vesicular transport and axonal trafficking, influencing the establishment and maintenance of synaptic connections. Altered expression or mutations in CLSTN2 have been linked to neurological disorders, including Alzheimer's disease, suggesting its significance in maintaining neural circuit stability and memory processes. Current research continues to explore CLSTN2's functional roles in neurobiology, with potential therapeutic implications for enhancing synaptic function and addressing neurodegenerative conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLSTN2
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene CLSTN2
Gene Accession No. Q9H4D0
Gene Alias 2900042C18Rik; AI448973; alcadein gamma; alcadein-gamma; alcagamma; alc-gamma; cadherin-related family member 13; calsyntenin 2; calsyntenin-2; CDHR13; Clstn2; CS2; CS2alcadein-gamma; Cst-2; Cstn2; FLJ39113; FLJ39499; Kiaa4134; MGC119560; mKIAA4134
Gene Symbols CLSTN2
Host Species Rabbit
Immunogen Recombinant Human Calsyntenin-2 protein (473-673AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64084
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.