Learn More
Invitrogen™ CLSTN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590686
Description
Immunogen sequence: SLQQRGLELT NTSAYLTIAG VESITVYEEI LRQARYRLRH GAALYTRKFR LSCSEMNGRY SSNEFIVEVN VLHSMNRVAH PSHVLSSQQF LHRGHQPPPE MAGHSLASSH RNSMIPSAAT L; Positive Samples: Mouse brain; Cellular Location: Cell membrane, Endoplasmic reticulum membrane, Golgi apparatus membrane, Single-pass type I membrane protein.

Specifications
CLSTN3 | |
Polyclonal | |
Unconjugated | |
Clstn3 | |
alcadein beta; alcadein-beta; alcbeta; Alc-beta; cadherin-related family member 14; calsyntenin 3; calsyntenin-3; CDHR14; CLSTN3; Cs3; Cst-3; Cstn3; KIAA0726; KIAA0726 MGC131797; MGC138488; mKIAA0726 | |
Rabbit | |
Affinity chromatography | |
RUO | |
232370 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.99 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q99JH7 | |
Clstn3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 730-850 of human CLSTN3 (NP_0555332). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.