Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLTA/CLTB Mouse anti-Human, Clone: CLC/1421, Abnova™

Mouse Monoclonal Antibody

Supplier:  Abnova Corporation MAB14481

Catalog No. 89-034-564


Only null left
Add to Cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human CLTA/CLTB.

  • High specificity
  • High reactivity
Specifications

Specifications

CLTA/CLTB
Monoclonal
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined b
P09496;P09497
CLTA
Recombinant protein corresponding to N-terminus of human CLTA/CLTB.
100 μg
Primary
Human
Liquid
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
CLC/1421
Mouse monoclonal antibody raised against partial recombinant human CLTA/CLTB.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
LCA
Mouse
Protein A/G purification
RUO
1211/1212
Store at 4°C.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only (RUO)