Learn More
CLTA Mouse anti-Human, Clone: OTI2F7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809615

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.Specifications
CLTA | |
Monoclonal | |
Unconjugated | |
CLTA | |
LCA | |
Mouse | |
Affinity Chromatography | |
RUO | |
1211 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2F7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P09496 | |
CLTA | |
Full length human recombit protein of human CLTA produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.