Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLTA Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | CLTA |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119026
|
Invitrogen™
PA5119026 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122093
|
Invitrogen™
PA5122093 |
20 μL |
Each for $237.00
|
|
|||||
Description
CLTA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.Specifications
CLTA | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
CLTA | |
AV026556; Clathrin light chain A; clathrin, light chain (Lca); clathrin, light chain A; clathrin, light polypeptide (Lca); clathryn light chain (LCA2); Clta; Lca; LCA1; LCA2; LCA3 | |
CLTA | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O08585 | |
12757 | |
Recombinant protein Clathrin, Light Polypeptide A. The antigen corresponds to amino acid range 5-234 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title