Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLTA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726656 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6656 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6656 Supplier Proteintech Group Inc Supplier No. 108521AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Clathrin consists of three heavy chain (192 kDa) and three light chains (LCa or LCb, 25-29 kDa) and self-assembles into a polyhedral cytoskeletal element. Self-assembly facilitates clathrin′s central role in membrane sorting and selective transport within the cell.

Specifications

Antigen CLTA
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CLTA
Gene Accession No. O08585, P08081, P09496
Gene Alias Clathrin light chain A, clathrin, light chain (Lca), CLTA, LCA
Gene Symbols Clta
Host Species Rabbit
Immunogen CLTA Fusion Protein Ag1299
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1211, 12757, 83800
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.