Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLTB Mouse anti-Human, Porcine, Clone: 1F9C2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CLTB |
---|---|
Clone | 1F9C2 |
Concentration | 1.32 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6703
![]() ![]() |
Proteintech Group Inc
662701IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.Specifications
CLTB | |
1.32 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Clathrin light chain B, clathrin, light chain (Lcb), CLTB, LCB | |
CLTB | |
IgG1 | |
Protein G | |
Antibody |
1F9C2 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
P09497 | |
100525860, 1212 | |
CLTB Fusion Protein Ag24347 | |
Primary | |
-20°C | |
CLTB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title