Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLTB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 104551AP150UL

Description
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.Specifications
CLTB | |
Polyclonal | |
Unconjugated | |
CLTB | |
Clathrin light chain B, clathrin, light chain (Lcb), CLTB, LCB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116561, 1212, 74325 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08082, P09497, Q6IRU5 | |
Cltb | |
CLTB Fusion Protein Ag0707 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction