Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLTC Mouse anti-Human, Clone: CLTC/1431, Abnova™

Catalog No. 89034566 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-566 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-566 Supplier Abnova Corporation Supplier No. MAB14483
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human CLTC.

  • High specificity
  • High reactivity

Specifications

Antigen CLTC
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone CLTC/1431
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human CLTC.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. Q00610;P53675
Gene Alias CHC/CHC17/CLH-17/CLTCL2/Hc/KIAA0034
Gene Symbols CLTC
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminus of human CLTC.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1213
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.