Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLTCL1 Mouse anti-Human, Clone: 1B5A10, Proteintech
SDP

Catalog No. 501736706 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6706 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6706 Supplier Proteintech Group Inc Supplier No. 662741IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody is specific to CLTCL1. It has no cross reaction to CLTC.

Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.

Specifications

Antigen CLTCL1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B5A10
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLTCL1
Gene Accession No. P53675
Gene Alias CHC22, Clathrin heavy chain 2, clathrin, heavy chain like 1, CLH 22, CLH22, CLTCL, CLTCL1, CLTD
Gene Symbols CLTCL1
Host Species Mouse
Immunogen Peptide
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8218
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.