Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLTCL1 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547886
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant human CHC17 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
Specifications
CLTCL1 | |
Polyclonal | |
Unconjugated | |
CLTCL1 | |
CHC22; clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 22; clathrin, heavy chain-like 1; Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; CLH22; CLH-22; CLTCL; CLTCL1; CLTD | |
Sheep | |
Antigen Affinity Chromatography | |
RUO | |
8218 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P53675 | |
CLTCL1 | |
E. coli-derived recombinant human Clathrin Heavy Chain 2/CHC22 Trp1521-Glu1640. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction