Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLTCL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568407 Supplier Invitrogen™ Supplier No. PA568407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLTCL1 Polyclonal Antibody for IHC (P)

Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLTCL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CLTCL1
Gene Accession No. P53675
Gene Alias CHC22; clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 22; clathrin, heavy chain-like 1; Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; CLH22; CLH-22; CLTCL; CLTCL1; CLTD
Gene Symbols CLTCL1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human CLTCL1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8218
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.