Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLUAP1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306085MFV500
Description
CLUAP1 Polyclonal antibody specifically detects CLUAP1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CLUAP1 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
clusterin associated protein 1, clusterin-associated protein 1, flagellar associated protein 22, qilin-like protein, homolog, FLJ13297, KIAA0643FAP22 | |
Produced in rabbits immunized with E. coli-derived Human CLUAP1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
23059 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction