Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLUAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528519 Supplier Invitrogen™ Supplier No. PA528519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116. Predicted reactivity: Mouse (91%), Rat (92%), Pig (90%), Chicken (81%), Chimpanzee (99%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CLUAP1 may play a role in cell proliferation or apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLUAP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Cluap1
Gene Accession No. Q96AJ1
Gene Alias 2310030D15Rik; 2610111M03Rik; CFAP22; cilia and flagella associated protein 22; Cluap1; clusterin associated protein 1; clusterin-associated protein 1; FAP22; flagellar associated protein 22, qilin-like protein, homolog; IFT38; KIAA0643; Qilin
Gene Symbols Cluap1
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 1 and 233 of Human CLUAP1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23059
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.