Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLUAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557893
Description
Immunogen sequence: VLYNAMKTKG MEGSEIVEED VNKFKFDLGS KIADLKAARQ LASEITSKGA SLYDLLGMEV ELREMRTEAI ARPLEINETE KVMRIAIKEI LTQVQKTKDL L Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
CLUAP1 may play a role in cell proliferation or apoptosis.
Specifications
CLUAP1 | |
Polyclonal | |
Unconjugated | |
Cluap1 | |
2310030D15Rik; 2610111M03Rik; CFAP22; cilia and flagella associated protein 22; Cluap1; clusterin associated protein 1; clusterin-associated protein 1; FAP22; flagellar associated protein 22, qilin-like protein, homolog; IFT38; KIAA0643; Qilin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23059 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96AJ1 | |
Cluap1 | |
Recombinant protein corresponding to Human CLUAP1. Recombinant protein control fragment (Product #RP-92456). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction